(4-tert-Butylphenoxy)acetic acid 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2-(4-(tert-Butyl)phenoxy)acetic acid

A17017-06 A17017-14 A17017-22
CAAAA17017-06EA 30.02 CAD
CAAAA17017-06 CAAAA17017-14 CAAAA17017-22
(4-tert-Butylphenoxy)acetic acid 98%
(4-tert-Butylphenoxy)acetic acid

Purity: 98% CAS No.: 1798-04-5 Molecular Formula: C12H16O3 Formula Weight: 208.26 MDL No.: MFCD00021758

Formula: (C₆H₄(C(CH₃)₃))CH₂COOH
MW: 208.26
Storage Temperature: Ambient
MDL Number: MFCD00021758
CAS Number: 1798-04-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR