Butyl benzoate 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A17010-0B
A17010-30
CAAAA17010-0BEA
107.12
CAD
CAAAA17010-0B
CAAAA17010-30
Butyl benzoate 98%
Butyl benzoate
Purity: 98%
CAS No.: 136-60-7
Molecular Formula: C11H14O2
Formula Weight: 178.23
Flash Point: 106°(222°F)
Refractive Index: 1.4980
Melting Point: -22°
Boiling Point: 250°
Beilstein Reference: 1867073
Merck Reference: 14,1552
MDL No.: MFCD00009439
Formula:
C₁₁H₁₄O₂ Boiling Pt: 250 °C Melting Pt: –22 °C Density: 1.011 Flash Pt: 106 °C(222 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00009439 CAS Number: 136-60-7 EINECS: 205-252-7 Merck Index: 14,1552 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...