Sodium trichloroacetate 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A17004-0B
A17004-0I
A17004-30
CAAAA17004-0BEA
154.37
CAD
CAAAA17004-0B
CAAAA17004-0I
CAAAA17004-30
Sodium trichloroacetate 97%
Sodium trichloroacetate
Purity: 97%
CAS No.: 650-51-1
Molecular Formula: CCl3CO2Na
Formula Weight: 185.37
Melting Point:
Formula:
C₂Cl₃NaO₂ Melting Pt: >300 °C Storage Temperature: Ambient |
MDL Number:
MFCD00064198 CAS Number: 650-51-1 EINECS: 211-479-2 UN: 3077 ADR: 9,III Merck Index: 14,9627 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...