2-Ethylbutyric acid 98%

Supplier: Thermo Scientific Chemicals
Danger

A16989-AK A16989-AU
CAAAA16989-AKEA 84.21 CAD
CAAAA16989-AK CAAAA16989-AU
2-Ethylbutyric acid 98%
2-Ethylbutyric acid

Purity: 98% CAS No.: 88-09-5 Molecular Formula: (CH3CH2)2CHCO2H Formula Weight: 116.16 Flash Point: 87°(188°F) Refractive Index: 1.4130 Melting Point: -15° Boiling Point: 192-194° Beilstein Reference: 1098634 Merck Reference: 14,3110 UN No.: 2922 MDL No.: MFCD00002670

Formula: C₆H₁₂O₂
Boiling Pt: 192…194 °C
Melting Pt: –15 °C
Density: 0.924
Flash Pt: 87 °C (188 °F)
Storage Temperature: Ambient
MDL Number: MFCD00002670
CAS Number: 88-09-5
EINECS: 201-796-4
UN: 2922
ADR: 8,III
Merck Index: 14,3110

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR