3,5-Dinitrosalicylic acid 97+%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2-Hydroxy-3,5-dinitrobenzoic acid, 3,5-Dinitro-2-hydroxybenzoic acid

A16988-0B A16988-18 A16988-30
CAAAAA16988-0BEA 899.02 CAD
CAAAAA16988-0B CAAAA16988-18 CAAAA16988-30
3,5-Dinitrosalicylic acid 97+%
3,5-Dinitrosalicylic acid

Purity: 97+% CAS No.: 609-99-4 Molecular Formula: C7H4N2O7 Formula Weight: 228.12 Melting Point: 169-174° Beilstein Reference: 2220661

Formula: C₇H₄N₂O₇
MW: 228.12 g/mol
Melting Pt: 169…174 °C
Storage Temperature: Ambient
MDL Number: MFCD00007104
CAS Number: 609-99-4
EINECS: 210-204-3
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR