(±)-2-Octanol 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A16985-0F
A16985-AE
A16985-AP
CAAAA16985-0FEA
195.92
CAD
CAAAA16985-0F
CAAAA16985-AE
CAAAA16985-AP
(±)-2-Octanol 98%
(±)-2-Octanol
Purity: 98%
CAS No.: 123-96-6
Molecular Formula: C8H18O
Formula Weight: 130.23
Flash Point: 76°(169°F)
Refractive Index: 1.4260
Odor: Pleasant
Melting Point: -38°
Boiling Point: 179-180°
Beilstein Reference: 1719325
Merck Reference: 14,6752
MDL No.: MFCD00004591
Formula:
C₈H₁₈O Boiling Pt: 179…180 °C Melting Pt: –38 °C Density: 0.819 Flash Pt: 76 °C(169 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00004591 CAS Number: 123-96-6 EINECS: 204-667-0 UN: 1987 ADR: 3,III Merck Index: 14,6752 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...