3-(Trifluoromethyl)cinnamic acid predominantly trans 98+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A16983-06
A16983-14
CAAAA16983-06EA
71.89
CAD
CAAAA16983-06
CAAAA16983-14
3-(Trifluoromethyl)cinnamic acid predominantly trans 98+%
3-(Trifluoromethyl)cinnamic acid
Purity: 98+%
CAS No.: 779-89-5
Molecular Formula: C10H7F3O2
Formula Weight: 216.16
Odor: Odorless
Melting Point: 132-136°
Beilstein Reference: 1963372
MDL No.: MFCD00004393
Formula:
C₁₀H₇F₃O₂ Boiling Pt: 292 °C (1013 hPa) Melting Pt: 132…136 °C Storage Temperature: Ambient |
MDL Number:
MFCD00004393 CAS Number: 779-89-5 EINECS: 212-301-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...