N-(3-(Trimethoxysilyl)propyl)ethylenediamine tech. 90%, Technical Grade
Supplier: Thermo Scientific Chemicals
|
Danger
|
A16981-0B
A16981-18
A16981-30
CAAAA16981-0BEA
334.96
CAD
CAAAA16981-0B
CAAAA16981-18
CAAAA16981-30
N-(3-(Trimethoxysilyl)propyl)ethylenediamine tech. 90%, Technical Grade
N-(3-(Trimethoxysilyl)propyl)ethylenediamine
Purity: 90%
CAS No.: 1760-24-3
Molecular Formula: (CH3O)3Si(CH2)3NHCH2CH2NH2
Formula Weight: 222.36
Flash Point: 136°(276°F)
Refractive Index: 1.4420
Boiling Point: 261-263°
Beilstein Reference: 636230
UN No.: 2735
MDL No.: MFCD00008173
Formula:
H₂NCH₂CH₂NHCH₂CH₂CH₂Si(OCH₃)₃ Boiling Pt: 261…263 °C Melting Pt: ∼ 0 °C Density: 1.010 Flash Pt: 136 °C(276 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00008173 CAS Number: 1760-24-3 EINECS: 217-164-6 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...