3-Heptanone 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A16977-AK
A16977-AU
CAAAA16977-AKEA
143.62
CAD
CAAAA16977-AK
CAAAA16977-AU
3-Heptanone 98%
3-Heptanone
Purity: 98%
CAS No.: 106-35-4
Molecular Formula: C7H14O
Formula Weight: 114.19
Flash Point: 41°(105°F)
Refractive Index: 1.4090
Odor: Pungent
Melting Point: -37°
Boiling Point: 147-149°
Beilstein Reference: 506161
UN No.: 1224
MDL No.: MFCD00009483
Formula:
C₇H₁₄O Boiling Pt: 147…149 °C Melting Pt: –37 °C Density: 0.818 Flash Pt: 41 °C (105 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00009483 CAS Number: 106-35-4 EINECS: 203-388-1 UN: 1224 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...