Triammonium citrate 97+%

Supplier: Thermo Scientific Chemicals
Warning

A16973-22 A16973-36
CAAAA16973-22EA 90.53 CAD
CAAAA16973-22 CAAAA16973-36
Triammonium citrate 97+%
tri-Ammonium citrate

MDL: MFCD00036406 Soluble in water


Purity: 97+% CAS No.: 3458-72-8 Molecular Formula: C6H17N3O7 Formula Weight: 243.22 Physical Form: Crystalline powder Appearance: White Melting Point: ca 180° dec. Solubility: Soluble in water MDL No.: MFCD00036406

Formula: C₆H₁₇N₃O₇
Melting Pt: ∼ 180 °C (dec.)
Density: 1 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00036406
CAS Number: 3458-72-8
EINECS: 222-394-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR