2,3-Difluorophenol 98+%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A16971-03
A16971-06
CAAAA16971-03EA
33.02
CAD
CAAAA16971-03
CAAAA16971-06
2,3-Difluorophenol 98+%
2,3-Difluorophenol
Purity: 98+%
CAS No.: 6418-38-8
Molecular Formula: C6H4F2O
Formula Weight: 130.09
Flash Point: 56°(132°F)
Odor: Phenol-like
Boiling Point: 53-55°/25mm
Beilstein Reference: 2082265
UN No.: 2921
MDL No.: MFCD00010262
Formula:
C₆H₄F₂O Boiling Pt: 53…55 °C/25 mm Melting Pt: 34…38 °C Flash Pt: 56 °C (132 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00010262 CAS Number: 6418-38-8 UN: 2925 ADR: 4.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...