4-Isopropylanisole 95%
Supplier: Thermo Scientific Chemicals
Warning
|
A16970-14
A16970-22
CAAAAA16970-36EA
763.95
CAD
CAAAAA16970-36
CAAAA16970-14
CAAAA16970-22
4-Isopropylanisole 95%
4-Isopropylanisole
Purity: 95%
CAS No.: 4132-48-3
Molecular Formula: C10H14O
Formula Weight: 150.22
Flash Point: 83°(181°F)
Refractive Index: 1.5030
Boiling Point: 212-213°
Beilstein Reference: 2080269
MDL No.: MFCD00015033
Formula:
C₁₀H₁₄O Boiling Pt: 212…213 °C Density: 0.935 Flash Pt: 83 °C(181 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00015033 CAS Number: 4132-48-3 EINECS: 223-952-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...