Xylitol 99%

Supplier: Thermo Scientific Chemicals

Synonyms: (2R,3r,4S)-Pentane-1,2,3,4,5-pentaol

A16944-22 A16944-36
CAAAA16944-22EA 64.62 CAD
CAAAA16944-22 CAAAA16944-36
Xylitol 99%
Xylitol

Purity: 99% CAS No.: 87-99-0 Molecular Formula: C5H12O5 Formula Weight: 152.15 Odor: Odorless Melting Point: 92-96° Beilstein Reference: 1720523 Merck Reference: 14,10085 MDL No.: MFCD00064292

Formula: C₅H₁₂O₅
MW: 152.15 g/mol
Boiling Pt: 495
Melting Pt: 94
Density: 1.515
Storage Temperature: Ambient
MDL Number: MFCD00064292
CAS Number: 87-99-0
EINECS: 201-788-0
Merck Index: 14,10085

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR