3-Nitrophenyl isocyanate 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A16920-06
A16920-14
CAAAA16920-06EA
78.53
CAD
CAAAA16920-06
CAAAA16920-14
3-Nitrophenyl isocyanate 97%
3-Nitrophenyl isocyanate
Purity: 97%
CAS No.: 3320-87-4
Molecular Formula: C7H4N2O3
Formula Weight: 164.12
Storage Temperature: 2°C to 8°C
Flash Point:
Formula:
C₇H₄N₂O₃ Boiling Pt: 130…131 °C/11 mm Melting Pt: 51…53 °C Flash Pt: >110 °C (230 °F) Storage Temperature: Refrigerator |
MDL Number:
MFCD00007220 CAS Number: 3320-87-4 EINECS: 222-025-8 UN: 2811 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...