3,5-Difluorobenzaldehyde 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A16917-03
A16917-06
A16917-14
CAAAAA16917-03EA
48.82
CAD
CAAAAA16917-03
CAAAA16917-06
CAAAA16917-14
3,5-Difluorobenzaldehyde 97%
3,5-Difluorobenzaldehyde
Purity: 97%
CAS No.: 32085-88-4
Molecular Formula: C7H4F2O
Formula Weight: 142.11
Flash Point: 56°(132°F)
Refractive Index: 1.4930
Boiling Point: 61-63°/43mm
Beilstein Reference: 2573392
UN No.: 1989
MDL No.: MFCD00010329
Formula:
C₇H₄F₂O Boiling Pt: 61…63 °C/43 mm Flash Pt: 56 °C (132 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00010329 CAS Number: 32085-88-4 UN: 1989 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...