2-Fluorotoluene 99+%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
1-Fluoro-2-methylbenzene, o-Fluorotoluene
A16912-0E
A16912-36
CAAAAA16912-0EEA
606.72
CAD
CAAAAA16912-0E
CAAAA16912-36
2-Fluorotoluene 99+%
2-Fluorotoluene
Purity: 99+%
CAS No.: 95-52-3
Molecular Formula: C7H7F
Formula Weight: 110.13
Flash Point: 12°(53°F)
Refractive Index: 1.4730
Odor: Aromatic
Melting Point: -87°
Boiling Point: 113-114°
Beilstein Reference: 1853362
Merck Reference: 14,4180
UN No.: 2388
MDL No.: MFCD00000322
Formula:
C₇H₇F MW: 110.13 g/mol Boiling Pt: 113…114 °C Melting Pt: –87 °C Density: 1.002 Flash Pt: 12 °C (53 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00000322 CAS Number: 95-52-3 EINECS: 202-428-5 UN: 2388 ADR: 3,II Merck Index: 14,4180 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...