(2-Bromophenyl)acetic acid 98+%

Supplier: Thermo Scientific Chemicals
Warning

A16900-22 A16900-06 A16900-14
CAAAAA16900-22EA 492.96 CAD
CAAAAA16900-22 CAAAA16900-06 CAAAA16900-14
(2-Bromophenyl)acetic acid 98+%
(2-Bromophenyl)acetic acid

Purity: 98+% CAS No.: 18698-97-0 Molecular Formula: C8H7BrO2 Formula Weight: 215.05 Melting Point: 105-106° Beilstein Reference: 2250655

Formula: (C₆H₄Br)CH₂COOH
Boiling Pt: 308 °C (1013 hPa)
Melting Pt: 105…106 °C
Storage Temperature: Ambient
MDL Number: MFCD00004314
CAS Number: 18698-97-0
EINECS: 242-509-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR