α'-Bromo-α,α,α-trifluoro-p-xylene 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 4-(Bromomethyl)benzotrifluoride, 1-(Bromomethyl)-4-(trifluoromethyl)benzene, 4-(Trifluoromethyl)benzyl bromide

A16895-06 A16895-14
CAAAA16895-06EA 118.66 CAD
CAAAA16895-06 CAAAA16895-14
α'-Bromo-α,α,α-trifluoro-p-xylene 98%
α'-Bromo-α,α,α-trifluoro-p-xylene

Purity: 98% CAS No.: 402-49-3 Molecular Formula: C8H6BrF3 Formula Weight: 239.04 Flash Point: 88°(190°F) Refractive Index: 1.4915 Odor: Pungent Melting Point: 29-33° Boiling Point: 65-66°/5mm Beilstein Reference: 638980 UN No.: 3261 MDL No.: MFCD00000403

Formula: C₈H₆BrF₃
MW: 239.03 g/mol
Boiling Pt: 65…66 °C/5 mm
Melting Pt: 29…33 °C
Density: 1.546
Flash Pt: 88 °C (190 °F)
Storage Temperature: Ambient
MDL Number: MFCD00000403
CAS Number: 402-49-3
EINECS: 206-947-8
UN: 3261
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR