2'-Fluoro-4'-methoxyacetophenone 99%

Supplier: Thermo Scientific Chemicals
Warning

A16891-06 A16891-14 A16891-22
CAAAA16891-06EA 21.01 CAD
CAAAA16891-06 CAAAA16891-14 CAAAA16891-22
2'-Fluoro-4'-methoxyacetophenone 99%
2'-Fluoro-4'-methoxyacetophenone

Purity: 99% CAS No.: 74457-86-6 Molecular Formula: C9H9FO2 Formula Weight: 168.17 Flash Point: 110°(230°F) Melting Point: 50-53° Boiling Point: 80-85°/0.1mm Beilstein Reference: 3539992 MDL No.: MFCD00042290

Formula: C₉H₉FO₂
MW: 168.17 g/mol
Boiling Pt: 80…85 °C/0.1 mm
Melting Pt: 50…53 °C
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00042290
CAS Number: 74457-86-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR