3,5-Di-tert-butyl-4-hydroxybenzoic acid 98%

Supplier: Thermo Scientific Chemicals

A16880-09 A16880-18
CAAAA16880-09EA 67.15 CAD
CAAAA16880-09 CAAAA16880-18
3,5-Di-tert-butyl-4-hydroxybenzoic acid 98%
3,5-Di-tert-butyl-4-hydroxybenzoic acid

Purity: 98% CAS No.: 1421-49-4 Molecular Formula: C15H22O3 Formula Weight: 250.34 Melting Point: 206-209° Beilstein Reference: 2216948

Formula: C₁₅H₂₂O₃
MW: 250.34 g/mol
Boiling Pt: 374 °C (1013 hPa)
Melting Pt: 206…209 °C
Storage Temperature: Ambient
MDL Number: MFCD00008827
CAS Number: 1421-49-4
EINECS: 215-823-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR