4-Iodo-m-xylene 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2,4-Dimethyliodobenzene, 1-Iodo-2,4-dimethylbenzene, 2,4-Dimethyl-1-iodobenzene, 1,3-Dimethyl-4-iodobenzene

A16878-09 A16878-18
CAAAA16878-09EA 89.11 CAD
CAAAA16878-09 CAAAA16878-18
4-Iodo-m-xylene 98%
4-Iodo-m-xylene

Purity: 98% CAS No.: 4214-28-2 Molecular Formula: C8H9I Formula Weight: 232.07 Flash Point: 110°(230°F) Refractive Index: 1.5980 Boiling Point: 231-232° Beilstein Reference: 1422602 MDL No.: MFCD00013706

Formula: C₈H₉I
Boiling Pt: 231…232 °C
Density: 1.628
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00013706
CAS Number: 4214-28-2
EINECS: 224-141-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR