2-Naphthalenesulfonyl chloride 97%

Supplier: Thermo Scientific Chemicals
Danger

A16872-36 A16872-14 A16872-22
CAAAAA16872-36EA 1360.38 CAD
CAAAAA16872-36 CAAAA16872-14 CAAAA16872-22
2-Naphthalenesulfonyl chloride 97%
2-Naphthalenesulphonyl chloride

Purity: 97% CAS No.: 93-11-8 Molecular Formula: C10H7ClO2S Formula Weight: 226.68 Melting Point: 74-78° Boiling Point: 200-202°/13mm UN No.: 3261 MDL No.: MFCD00004087

Formula: C₁₀H₇ClO₂S
Boiling Pt: 200…202 °C/13 mm
Melting Pt: 74…78 °C
Storage Temperature: Ambient
MDL Number: MFCD00004087
CAS Number: 93-11-8
EINECS: 202-219-9
UN: 3261
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR