2,6-Naphthalenedicarboxylic acid 98+%

Supplier: Thermo Scientific Chemicals

A16868-06 A16868-14 A16868-22
CAAAA16868-06EA 91.8 CAD
CAAAA16868-06 CAAAA16868-14 CAAAA16868-22
2,6-Naphthalenedicarboxylic acid 98+%
2,6-Naphthalenedicarboxylic acid

Purity: 98+% CAS No.: 1141-38-4 Molecular Formula: C12H8O4 Formula Weight: 216.19 Odor: Odorless Melting Point: 300° Beilstein Reference: 2051257 MDL No.: MFCD00004105

Formula: C₁₂H₈O₄
Boiling Pt: 422 °C (1013 hPa)
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00004105
CAS Number: 1141-38-4
EINECS: 214-527-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR