2,6-Di-tert-butyl-p-cresol 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A16863-0B
A16863-30
CAAAA16863-0BEA
119.61
CAD
CAAAA16863-0B
CAAAA16863-30
2,6-Di-tert-butyl-p-cresol 99%
2,6-Di-tert-butyl-p-cresol
Antioxidant
Purity: 99%
CAS No.: 128-37-0
Molecular Formula: C15H24O
Formula Weight: 220.36
Flash Point: 127°(260°F)
Odor: Mild
Boiling Point: 264-265°
Beilstein Reference: 1911640
Merck Reference: 14,1548
UN No.: 3077
MDL No.: MFCD00011644
Formula:
C₁₅H₂₄O Boiling Pt: 264…265 °C Melting Pt: 69…72 °C Density: 1.048 Flash Pt: 127 °C (260 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00011644 CAS Number: 128-37-0 EINECS: 204-881-4 UN: 3077 ADR: 9,III Merck Index: 14,1548 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...