(1-Bromoethyl)benzene 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
1-Bromo-1-phenylethane, α-Methylbenzyl bromide
A16839-14
A16839-22
A16839-36
CAAAA16839-14EA
79.16
CAD
CAAAA16839-14
CAAAA16839-22
CAAAA16839-36
(1-Bromoethyl)benzene 97%
1-Bromo-1-phenylethane
Purity: 97%
CAS No.: 585-71-7
Molecular Formula: C8H9Br
Formula Weight: 185.06
Flash Point: 81°(177°F)
Refractive Index: 1.5600
Melting Point: -65°
Boiling Point: 200°
Beilstein Reference: 507210
MDL No.: MFCD00000139
Formula:
C₆H₅CHBrCH₃ Boiling Pt: 200 °C Melting Pt: –65 °C Density: 1.356 Flash Pt: 81 °C(177 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00000139 CAS Number: 585-71-7 EINECS: 209-560-2 UN: 3334 ADR: 8,II |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...