2,3,4,5,6-Pentafluorostyrene 98% stabilized
Supplier: Thermo Scientific Chemicals
|
Warning
|
Stabilization:
Stabilised with 4-tert-butylpyrocatechol (TBC) 250 ppm
A16833-06
A16833-14
CAAAA16833-06EA
121.98
CAD
CAAAA16833-06
CAAAA16833-14
2,3,4,5,6-Pentafluorostyrene 98% stabilized
2,3,4,5,6-Pentafluorostyrene
Purity: 98%
CAS No.: 653-34-9
Molecular Formula: C8H3F5
Formula Weight: 194.10
Storage Temperature: 2°C to 8°C
Flash Point: 34°(93°F)
Refractive Index: 1.4460
Odor: Characteristic
Boiling Point: 139-140°
Beilstein Reference: 1874390
UN No.: 1993
MDL No.: MFCD00000300
Formula:
C₈H₃F₅ MW: 194.1 g/mol Boiling Pt: 139…140 °C Density: 1.406 Flash Pt: 34 °C (93 °F) Storage Temperature: Refrigerator |
MDL Number:
MFCD00000300 CAS Number: 653-34-9 EINECS: 211-500-5 UN: 1993 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...