D-(+)-Glucose, anhydrous 99%
Supplier: Thermo Scientific Chemicals
Synonyms:
Corn sugar, ᴅ-(+)-Glucose, Glucose, D-(+)-Dextrose, (2R,3S,4R,5R)-2,3,4,5,6-Pentahydroxyhexanal, Glucolin, Dextrose, D-Glucose
A16828-0C
A16828-0E
A16828-36
CAAAAA16828-0CEA
891.12
CAD
CAAAAA16828-0C
CAAAA16828-0E
CAAAA16828-36
D-(+)-Glucose, anhydrous 99%
D(+)-Glucose
Purity: 99%
CAS No.: 50-99-7
Molecular Formula: C6H12O6
Formula Weight: 180.16
Physical Form: Cryst powder
Appearance: White
Melting Point: 147-153°
Beilstein Reference: 1281608
Merck Reference: 14,4459
Optical Rotation: +53° (c=10 in water, 3h)
MDL No.: MFCD00063774
Formula:
C₆H₁₂O₆ Melting Pt: 147…153 °C Density: 1.54 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00063774 CAS Number: 50-99-7 EINECS: 200-075-1 Merck Index: 14,4459 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...