4,4'-Diaminodiphenyl ether 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A16815-18
A16815-22
A16815-36
CAAAAA16815-18EA
60.04
CAD
CAAAAA16815-18
CAAAA16815-22
CAAAA16815-36
4,4'-Diaminodiphenyl ether 98%
4,4'-Diaminodiphenyl ether
Purity: 98%
CAS No.: 101-80-4
Molecular Formula: C12H12N2O
Formula Weight: 200.24
Flash Point: 218°(424°F)
Melting Point: 189-192°
Boiling Point: 190°/0.1mm
Beilstein Reference: 475735
UN No.: 3077
MDL No.: MFCD00007863
Formula:
C₁₂H₁₂N₂O Boiling Pt: 190 °C/0.1 mm Melting Pt: 189…192 °C Flash Pt: 218 °C (424 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00007863 CAS Number: 101-80-4 EINECS: 202-977-0 UN: 2811 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...