D-Gluconic acid 50% in aqueous solution
Supplier: Thermo Scientific Chemicals
A16806-0F
A16806-AP
CAAAA16806-0FEA
136.35
CAD
CAAAA16806-0F
CAAAA16806-AP
D-Gluconic acid 50% in aqueous solution
D-Gluconic acid
CAS No.: 526-95-4
Molecular Formula: C6H12O7
Formula Weight: 196.16(anhy)
Refractive Index: 1.4180
Beilstein Reference: 1726055
Merck Reference: 14,4456
Optical Rotation: +10° (neat)
UN No.: 3265
MDL No.: MFCD00004240
Formula:
HOH₂C(CH(OH))₄COOH Density: 1.234 Storage Temperature: Ambient |
MDL Number:
MFCD00004240 CAS Number: 526-95-4 EINECS: 208-401-4 UN: 3265 ADR: 8,III Merck Index: 14,4456 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...