4-Methoxyphenyl isocyanate 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
1-Isocyanato-4-methoxybenzene
A16805-06
A16805-14
CAAAA16805-06EA
83.11
CAD
CAAAA16805-06
CAAAA16805-14
CAAAA16805-22
4-Methoxyphenyl isocyanate 98%
4-Methoxyphenyl isocyanate
Purity: 98%
CAS No.: 5416-93-3
Molecular Formula: C8H7NO2
Formula Weight: 149.15
Storage Temperature: 2°C to 8°C
Flash Point: 98°(208°F)
Refractive Index: 1.5470
Odor: Pungent
Boiling Point: 119-120°/20mm
Beilstein Reference: 471920
UN No.: 2206
MDL No.: MFCD00002026
Formula:
C₈H₇NO₂ Boiling Pt: 119…120 °C/20 mm Density: 1.151 Flash Pt: 98 °C (208 °F) Storage Temperature: Refrigerator |
MDL Number:
MFCD00002026 CAS Number: 5416-93-3 EINECS: 226-513-1 UN: 2206 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...