Acetylthiocholine iodide 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A16802-03
A16802-06
A16802-14
CAAAA16802-03EA
46.61
CAD
CAAAA16802-03
CAAAA16802-06
CAAAA16802-14
Acetylthiocholine iodide 98%
Acetylthiocholine iodide
Purity: 98%
CAS No.: 1866-15-5
Molecular Formula: CH3COSCH2CH2N(CH3)3I
Formula Weight: 289.18
Storage Temperature: 2°C to 8°C
Melting Point: 205-209°
Beilstein Reference: 3916578
UN No.: 2811
MDL No.: MFCD00011819
Formula:
H₃CC(S)O(CH₂)₂N(CH₃)₃I Melting Pt: 205…209 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00011819 CAS Number: 1866-15-5 EINECS: 217-474-1 UN: 2811 ADR: 6.1,III |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...