α,α,α,α',α',α'-Hexafluoro-5-nitroxylene 97+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A16797-06
A16797-14
CAAAA16797-06EA
57.04
CAD
CAAAA16797-06
CAAAA16797-14
α,α,α,α',α',α'-Hexafluoro-5-nitroxylene 97+%
α,α,α,α',α',α'-Hexafluoro-5-nitroxylene
Purity: 97+%
CAS No.: 328-75-6
Molecular Formula: C8H3F6NO2
Formula Weight: 259.11
Flash Point: 77°(170°F)
Refractive Index: 1.4270
Boiling Point: 75-76°/11mm
Beilstein Reference: 1998251
MDL No.: MFCD00000384
Formula:
C₈H₃F₆NO₂ Boiling Pt: 75…76 °C (11 mmHg) Density: 1,535 Flash Pt: 77 °C (170 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00000384 CAS Number: 328-75-6 EINECS: 206-336-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...