2',3'-Difluoroacetophenone 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
1-(2,3-Difluorophenyl)ethan-1-one, 1-(2,3-Difluorophenyl)ethanone, 2,3-Difluoroacetophenone
A16786-14
A16786-06
CAAAAA16786-14EA
439.24
CAD
CAAAAA16786-14
CAAAA16786-03
CAAAA16786-06
2',3'-Difluoroacetophenone 97%
2',3'-Difluoroacetophenone
Purity: 97%
CAS No.: 18355-80-1
Molecular Formula: C8H6F2O
Formula Weight: 156.13
Refractive Index: 1.4890
Odor: Characteristic
Boiling Point: 84-85°/20mm
Beilstein Reference: 2963572
MDL No.: MFCD00061274
Formula:
C₈H₆F₂O MW: 156.13 g/mol Boiling Pt: 84…85 °C/20 mm Storage Temperature: Ambient |
MDL Number:
MFCD00061274 CAS Number: 18355-80-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...