4-Fluoro-1-naphthoic acid 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A16778-03
A16778-06
CAAAA16778-03EA
79.32
CAD
CAAAA16778-03
CAAAA16778-06
4-Fluoro-1-naphthoic acid 97%
4-Fluoro-1-naphthoic acid
Purity: 97%
CAS No.: 573-03-5
Molecular Formula: C11H7FO2
Formula Weight: 190.18
Melting Point: 223-226°
Beilstein Reference: 2257292
Formula:
C₁₁H₇FO₂ Melting Pt: 223…226 °C Storage Temperature: Ambient |
MDL Number:
MFCD00004011 CAS Number: 573-03-5 EINECS: 209-346-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...