α-Chloro-3-nitrotoluene 98%
Supplier: Thermo Scientific Chemicals
A16775-30
A16775-09
A16775-18
CAAAAA16775-30EA
690.46
CAD
CAAAAA16775-30
CAAAA16775-09
CAAAA16775-18
α-Chloro-3-nitrotoluene 98%
α-Chloro-3-nitrotoluene
Purity: 98%
CAS No.: 619-23-8
Molecular Formula: C7H6ClNO2
Formula Weight: 171.58
Flash Point:
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...