8-Aminoquinoline 98+%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Quinolin-8-ylamine

A16752-03 A16752-06
CAAAA16752-03EA 34.76 CAD
CAAAA16752-03 CAAAA16752-06
8-Aminoquinoline 98+%
8-Aminoquinoline

Purity: 98+% CAS No.: 578-66-5 Molecular Formula: C9H8N2 Formula Weight: 144.18 Melting Point: 63-67° Boiling Point: 174°/26mm MDL No.: MFCD00006809

Formula: C₉H₈N₂
MW: 144.18 g/mol
Boiling Pt: 290
Melting Pt: 64
Storage Temperature: Ambient
MDL Number: MFCD00006809
CAS Number: 578-66-5
EINECS: 209-427-9
UN: 2811
ADR: 6.1 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR