1,3,3-Trimethyl-2-methyleneindoline 95%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A16741-18
A16741-30
CAAAA16741-18EA
105.07
CAD
CAAAA16741-18
CAAAA16741-30
1,3,3-Trimethyl-2-methyleneindoline 95%
1,3,3-Trimethyl-2-methyleneindoline
Purity: 95%
CAS No.: 118-12-7
Molecular Formula: C12H15N
Formula Weight: 173.26
Flash Point: 101°(213°F)
Refractive Index: 1.5780
Melting Point: -10°
Boiling Point: 247-248°
Beilstein Reference: 131162
MDL No.: MFCD00005813
Formula:
C₁₂H₁₅N Boiling Pt: 247…248 °C Melting Pt: –10 °C Density: 0.979 Flash Pt: 101 °C(213 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00005813 CAS Number: 118-12-7 EINECS: 204-235-1 UN: 3082 ADR: 9,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...