3-(Trifluoromethyl)benzyl alcohol 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A16739-06
A16739-14
CAAAA16739-06EA
78.53
CAD
CAAAA16739-06
CAAAA16739-14
3-(Trifluoromethyl)benzyl alcohol 97%
3-(Trifluoromethyl)benzyl alcohol
Purity: 97%
CAS No.: 349-75-7
Molecular Formula: C8H7F3O
Formula Weight: 176.14
Flash Point: 84°(183°F)
Refractive Index: 1.4600
Boiling Point: 79-80°/3mm
Beilstein Reference: 1450161
MDL No.: MFCD00004645
Formula:
C₈H₇F₃O Boiling Pt: 79…80 °C/3 mm Density: 1,293 Flash Pt: 84 °C(183 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00004645 CAS Number: 349-75-7 EINECS: 206-489-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...