α,α,α-Trifluoro-m-toluic acid 98+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A16737-09
A16737-18
CAAAAA16737-30EA
2827.58
CAD
CAAAAA16737-30
CAAAA16737-09
CAAAA16737-18
α,α,α-Trifluoro-m-toluic acid 98+%
α,α,α-Trifluoro-m-toluic acid
Purity: 98+%
CAS No.: 454-92-2
Molecular Formula: C8H5F3O2
Formula Weight: 190.12
Melting Point: 103-107°
Boiling Point: 237-240°
MDL No.: MFCD00002519
Formula:
C₈H₅F₃O₂ Boiling Pt: 237…240 °C Melting Pt: 103…107 °C Density: 1.371 g/cm³ (25 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00002519 CAS Number: 454-92-2 EINECS: 207-230-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...