2',6'-Dimethoxyacetophenone 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 1-(2,6-Dimethoxyphenyl)ethan-1-one, 1-(2,6-Dimethoxyphenyl)ethanone

A16733-06 A16733-14
CAAAA16733-06EA 100.01 CAD
CAAAA16733-06 CAAAA16733-14 CAAAA16733-22
2',6'-Dimethoxyacetophenone 98%
2',6'-Dimethoxyacetophenone

Purity: 98% CAS No.: 2040-04-2 Molecular Formula: C10H12O3 Formula Weight: 180.20 Melting Point: 68-71° Boiling Point: 135-136°/2mm MDL No.: MFCD00008729

Formula: C₁₀H₁₂O₃
MW: 180.2
Boiling Pt: 314
Melting Pt: 68…71 °C
Storage Temperature: Ambient
MDL Number: MFCD00008729
CAS Number: 2040-04-2
EINECS: 218-034-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR