3,5-Dinitrobenzoyl chloride 98+%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A16729-14
A16729-22
CAAAA16729-14EA
70.47
CAD
CAAAA16729-14
CAAAA16729-22
3,5-Dinitrobenzoyl chloride 98+%
3,5-Dinitrobenzoyl chloride
Purity: 98+%
CAS No.: 99-33-2
Molecular Formula: C7H3ClN2O5
Formula Weight: 230.57
Melting Point: 67-70°
Boiling Point: 196°/12mm
Merck Reference: 14,3277
UN No.: 3261
MDL No.: MFCD00007248
Formula:
C₇H₃ClN₂O₅ MW: 230.56 g/mol Boiling Pt: 196 °C (12 mmHg) Melting Pt: 67…70 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00007248 CAS Number: 99-33-2 EINECS: 202-750-6 UN: 3261 ADR: 8 III Merck Index: 14,3277 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...