(2,3-Difluorophenyl)acetic acid 98%

Supplier: Thermo Scientific Chemicals
Warning

A16728-03 A16728-06
CAAAAA16728-14EA 966.38 CAD
CAAAAA16728-14 CAAAA16728-03 CAAAA16728-06
(2,3-Difluorophenyl)acetic acid 98%
(2,3-Difluorophenyl)acetic acid

Purity: 98% CAS No.: 145689-41-4 Molecular Formula: C8H6F2O2 Formula Weight: 172.13 Melting Point: 117-119° MDL No.: MFCD00040968

Formula: (C₆H₃F₂)CH₂COOH
Melting Pt: 117…119 °C
Storage Temperature: Ambient
MDL Number: MFCD00040968
CAS Number: 145689-41-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR