1-Pentene 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
alpha-n-Amylene, Propylethylene
A16723-14
A16723-22
CAAAA16723-14EA
103.33
CAD
CAAAA16723-14
CAAAA16723-22
1-Pentene 97%
1-Pentene
Purity: 97%
CAS No.: 109-67-1
Molecular Formula: CH3(CH2)2CH=CH2
Formula Weight: 70.14
Storage Temperature: 2°C to 8°C
Flash Point: -51°(-60°F)
Refractive Index: 1.3715
Melting Point: -138°
Boiling Point: 30-31°
Beilstein Reference: 1731629
Merck Reference: 14,7122
UN No.: 1108
MDL No.: MFCD00003567
Formula:
C₅H₁₀ MW: 70.13 g/mol Boiling Pt: 30…31 °C Melting Pt: –138 °C Density: 0.640 Flash Pt: –51 °C (–60 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00003567 CAS Number: 109-67-1 EINECS: 203-694-5 UN: 1108 ADR: 3,I Merck Index: 14,7122 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...