2,3-Difluorobenzoic acid 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A16717-06
A16717-14
CAAAA16717-03EA
67.59
CAD
CAAAA16717-03
CAAAA16717-06
CAAAA16717-14
2,3-Difluorobenzoic acid 98%
2,3-Difluorobenzoic acid
Purity: 98%
CAS No.: 4519-39-5
Molecular Formula: C7H4F2O2
Formula Weight: 158.11
Odor: Odorless
Melting Point: 160-164°
Beilstein Reference: 2640781
MDL No.: MFCD00010267
Formula:
C₇H₄F₂O₂ Melting Pt: 160…164 °C Storage Temperature: Ambient |
MDL Number:
MFCD00010267 CAS Number: 4519-39-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...