Alizarin complexone, (max. 10% H₂O)
Supplier: Thermo Scientific Chemicals
A16699-03
A16699-06
CAAAA16699-03EA
149.47
CAD
CAAAA16699-03
CAAAA16699-06
Alizarin complexone, (max. 10% H₂O)
Alizarin complexone
CAS No.: 3952-78-1
Molecular Formula: C19H15NO8
Formula Weight: 385.33
Melting Point: 180-182°
Beilstein Reference: 2190028
MDL No.: MFCD00001202
Formula:
C₁₉H₁₅NO₈ Melting Pt: 180…182 °C Storage Temperature: Ambient |
MDL Number:
MFCD00001202 CAS Number: 3952-78-1 EINECS: 223-544-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...