3-Benzoylpyridine 98+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Phenyl 3-pyridyl ketone, Phenyl(pyridin-3-yl)methanone
A16695-22
A16695-06
A16695-14
CAAAAA16695-22EA
485.06
CAD
CAAAAA16695-22
CAAAA16695-06
CAAAA16695-14
3-Benzoylpyridine 98+%
3-Benzoylpyridine
Purity: 98+%
CAS No.: 5424-19-1
Molecular Formula: C12H9NO
Formula Weight: 183.21
Flash Point: 150°(302°F)
Melting Point: 36-40°
Boiling Point: 307°
Beilstein Reference: 120234
MDL No.: MFCD00006394
Formula:
C₁₂H₉NO MW: 183.21 g/mol Boiling Pt: 307 °C Melting Pt: 36…40 °C Flash Pt: 150 °C (302 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00006394 CAS Number: 5424-19-1 EINECS: 226-561-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...