Agarose ME for electrophoresis

Supplier: Thermo Scientific Chemicals

A16693-06 A16693-14
CAAAA16693-06EA 71.1 CAD
CAAAA16693-06 CAAAA16693-14
Agarose ME for electrophoresis
Agarose

CAS No.: 9012-36-6 MDL No.: MFCD00081294

Melting Pt: 60…90 °C
Storage Temperature: Ambient
MDL Number: MFCD00081294
CAS Number: 9012-36-6
EINECS: 232-731-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=526c20a29610acf6900f898ff245c5602558c785&vid=ae34e4c76a4d7838d4dc9501397f6741b1b97d5e&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR