2-Bromo-5-chlorothiophene 97%

Supplier: Thermo Scientific Chemicals
Warning

A16682-22 A16682-06 A16682-14
CAAAAA16682-22EA 464.52 CAD
CAAAAA16682-22 CAAAA16682-06 CAAAA16682-14
2-Bromo-5-chlorothiophene 97%
2-Bromo-5-chlorothiophene

Purity: 97% CAS No.: 2873-18-9 Molecular Formula: C4H2BrClS Formula Weight: 197.48 Flash Point: 85°(185°F) Refractive Index: 1.5960 Boiling Point: 69-70°/18mm Beilstein Reference: 109897 UN No.: 2810 MDL No.: MFCD00014523

Formula: C₄H₂BrClS
MW: 197.48 g/mol
Boiling Pt: 69…70 °C (18 mmHg)
Melting Pt: –2 °C
Density: 1.803
Flash Pt: 85 °C (185 °F)
Storage Temperature: Ambient
MDL Number: MFCD00014523
CAS Number: 2873-18-9
EINECS: 220-708-5
UN: 2810

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR