2,4-Dibromo-3-pentanone mixture of stereoisomers 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A16680-09
A16680-18
CAAAA16680-09EA
203.82
CAD
CAAAA16680-09
CAAAA16680-18
2,4-Dibromo-3-pentanone mixture of stereoisomers 97%
2,4-Dibromo-3-pentanone
Purity: 97%
CAS No.: 815-60-1
Molecular Formula: C5H8Br2O
Formula Weight: 243.91
Storage Temperature: 2°C to 8°C
Odor: Pungent
Boiling Point: 67-82°/10mm
UN No.: 3265
MDL No.: MFCD00015719
Formula:
H₃CBrCHC(O)BrCHCH₃ MW: 243.93 g/mol Boiling Pt: 67…82 °C/10 mm Storage Temperature: Ambient |
MDL Number:
MFCD00015719 CAS Number: 815-60-1 UN: 3265 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...