2'-Nitroacetanilide 98+%

Supplier: Thermo Scientific Chemicals
Warning

A16677-06 A16677-14
CAAAA16677-06EA 58.46 CAD
CAAAA16677-06 CAAAA16677-14 CAAAA16677-22
2'-Nitroacetanilide 98+%
2'-Nitroacetanilide

Purity: 98+% CAS No.: 552-32-9 Molecular Formula: C8H8N2O3 Formula Weight: 180.16 Melting Point: 90-93° Beilstein Reference: 1959178 Merck Reference: 14,6581 MDL No.: MFCD00016991

Formula: C₈H₈N₂O₃
Melting Pt: 90…93 °C
Density: 1.420
Storage Temperature: Ambient
MDL Number: MFCD00016991
CAS Number: 552-32-9
EINECS: 209-009-6
Merck Index: 14,6581

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR